5-Bromoacenaphthene ≥93.0% (by GC)
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synonyms:
1,2-Dihydro-5-bromoacenaphthylene, 5-Bromo-1,2-dihydroacenaphthylene
B1283-25G
B1283-5G
TCIAB1283-25GEA
0
GBP
TCIAB1283-25G
TCIAB1283-5G
5-Bromoacenaphthene ≥93.0% (by GC)
5-Bromoacenaphthene
Formula:
C₁₂H₉Br MW: 233.11 g/mol Boiling Pt: 335°C Melting Pt: 54 °C Density: 1,1452 g/cm³ (20 °C) |
MDL Number:
MFCD00003809 CAS Number: 2051-98-1 EINECS: 218-138-7 |
Specification Test Results
Appearance | White to Yellow powder to crystal |
Purity (GC) | min. 93.0 % |
Melting point | 53.0 to 57.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...