2',5'-Dimethylacetophenone ≥98.0% (by GC)

Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: 1-(2,5-Dimethylphenyl)ethanone, 1-(2,5-Dimethylphenyl)ethan-1-one

D1880-25ML D1880-250ML
TCIAD1880-25MLEA 0 GBP
TCIAD1880-25ML TCIAD1880-250ML
2',5'-Dimethylacetophenone ≥98.0% (by GC)
2',5'-Dimethylacetophenone
Formula: C₁₀H₁₂O
MW: 148.2 g/mol
Boiling Pt: 112°C
Melting Pt: –18 °C
Density: 1,0154 g/cm³ (0 °C)
Flash Pt: 96°C
MDL Number: MFCD00014988
CAS Number: 2142-73-6
EINECS: 218-399-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=8a8971d7133507be37eab8f84f2a11f279050a76&vid=8a8971d7133507be37eab8f84f2a11f279050a76&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance Colorless to Light yellow to Light orange clear liquid
Purity (GC) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR