2,4-Dimethoxybenzyl alcohol ≥98.0% (by GC)

Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

D1835-25G D1835-5G
TCIAD1835-25GEA 0 GBP
TCIAD1835-25G TCIAD1835-5G
2,4-Dimethoxybenzyl alcohol ≥98.0% (by GC)
2,4-Dimethoxybenzyl alcohol
Formula: C₉H₁₂O₃
MW: 168.19 g/mol
Boiling Pt: 135°C
Melting Pt: 34…35 °C
MDL Number: MFCD00004614
CAS Number: 7314-44-5
EINECS: 230-775-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=16a62edb74c3d8e6af170f7b8a103e2b7d960e22&vid=16a62edb74c3d8e6af170f7b8a103e2b7d960e22&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance White or Colorless to Almost white or Almost colorless powder to lump to clear liquid
Purity (GC) min. 98.0 %
Melting point 34.0 to 37.0 deg-C
Solubility in Methanol almost transparency

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR