3-Bromobenzenesulphonyl chloride ≥98.0% (by GC, titration analysis)
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Danger
|
B2389-1G
B2389-5G
TCIAB2389-1GEA
0
GBP
TCIAB2389-1G
TCIAB2389-5G
3-Bromobenzenesulphonyl chloride ≥98.0% (by GC, titration analysis)
3-Bromobenzenesulphonyl chloride
Formula:
C₆H₄BrClO₂S MW: 255.52 g/mol Boiling Pt: 95 - 95,5 °C (0.8 -rr) Melting Pt: 31…33 °C Density: 1,773 g/cm³ (20 °C) |
MDL Number:
MFCD00052313 CAS Number: 2905-24-0 UN: 3265 ADR: 8,II |
Specification Test Results
Purity (GC) | min. 98.0 % |
Purity (Argentmetric Titration) | min. 98.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...