2,2-Dimethylsuccinic acid ≥98.0% (by GC, titration analysis)

Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Warning

Synonyms: 2,2-Dimethyl-1,4-butanedioic acid, α,α-Dimethylsuccinic acid

D2184-25G D2184-5G
TCIAD2184-25GEA 0 GBP
TCIAD2184-25G TCIAD2184-5G
2,2-Dimethylsuccinic acid ≥98.0% (by GC, titration analysis)
2,2-Dimethylsuccinic acid
Formula: C₆H₁₀O₄
MW: 146.14 g/mol
Boiling Pt: 296 °C (1013 hPa)
Melting Pt: 143 °C
Density: 1,323 g/cm³ (20 °C)
MDL Number: MFCD00004196
CAS Number: 597-43-3
EINECS: 209-899-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d5bd786bbc6471c554b9fe5d30de30b40d0b8cca&vid=d5bd786bbc6471c554b9fe5d30de30b40d0b8cca&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Purity (Neutralization titration) min. 98.0 %
Melting point 140.0 to 143.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR