6-Bromo-1-hexanol ≥95.0% (by GC)

Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Warning

Synonyms: 6-Bromohexan-1-ol, Hexamethylene bromohydrin, 1-Bromo-6-hydroxyhexane

B1822-25G B1822-5G
TCIAB1822-25GEA 0 GBP
TCIAB1822-25G TCIAB1822-5G
6-Bromo-1-hexanol ≥95.0% (by GC)
6-Bromo-1-hexanol
Formula: BrH₂C(CH₂)₄CH₂OH
MW: 181.07 g/mol
Boiling Pt: 114°C
Density: 1,3838 g/cm³ (24 °C)
Storage Temperature: Refrigerator
MDL Number: MFCD00002983
CAS Number: 4286-55-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=91105080a1df94281774d72ca2656e496ebd9e54&vid=91105080a1df94281774d72ca2656e496ebd9e54&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance Colorless to Light yellow to Light orange clear liquid
Purity (GC) min. 95.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR