2,4-Dichloro-3,5-xylenol ≥98.0% (by GC)

Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Danger

Synonyms: 2,4-Dichloro-3,5-dimethylphenol

D1109-25G D1109-500G
TCIAD1109-25GEA 0 GBP
TCIAD1109-25G TCIAD1109-500G
2,4-Dichloro-3,5-xylenol ≥98.0% (by GC)
2,4-Dichloro-3,5-xylenol
Formula: C₈H₈Cl₂O
MW: 191.06 g/mol
Boiling Pt: 269 °C (1013 hPa)
Melting Pt: 96 °C
MDL Number: MFCD00019981
CAS Number: 133-53-9
EINECS: 205-109-9
UN: 2430
ADR: 8,III
Merck Index: 14,03076
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=97cc384359491b9118d274b95fcf1b056e3d6356&vid=97cc384359491b9118d274b95fcf1b056e3d6356&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance White to Light yellow to Light red powder to crystal
Purity (GC) min. 98.0 %
Melting point 92.0 to 96.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR