4-(1H-1,2,4-Triazol-1-yl)aniline

Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: 4-(1,2,4-Triazol-1-yl)aniline, 1-(4-Aminophenyl)-1,2,4-triazole

ST3522.1 ST3522.10
MENAST3522.1EA 0 GBP
MENAST3522.1 MENAST3522.10
4-(1H-1,2,4-Triazol-1-yl)aniline
4-(1H-1,2,4-Triazol-1-yl)aniline
Formula: C₈H₈N₄
MW: 160.1759 g/mol
MDL Number: MFCD00171470
CAS Number: 6523-49-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR