1-(2-Tetrahydropyranyl)-1H-pyrazole-5-boronic acid pinacol ester

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: 1-(Tetrahydropyran-2-yl)-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyrazole, 1-(Tetrahydro-2H-pyran-2-yl)-1H-pyrazole-5-boronic acid Pinacol ester, 1-(Tetrahydropyran-2-yl)-1H-pyrazole-5-boronic acid pinacol ester, 1-(Tetrahydro-2H-pyran-2-yl)-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-pyrazole

PN-8781-5G PN-8781-1G PN-8781-100G
COBBPN-8781-5GEA 0 GBP
COBBPN-8781-5G COBBPN-8781-1G COBBPN-8781-100G
1-(2-Tetrahydropyranyl)-1H-pyrazole-5-boronic acid pinacol ester
1-(2-Tetrahydropyranyl)-1H-pyrazole-5-boronic acid pinacol ester
Formula: C₁₄H₂₃BN₂O₃
MW: 278.16 g/mol
MDL Number: MFCD09037501
CAS Number: 903550-26-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR