(S)-(-)-α,α-Diphenylprolinol, Sigma-Aldrich®

Supplier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: (S)-(-)-α,α-Diphenyl-2-pyrrolidinemethanol

M-Clarity™ quality level = MQ200

368199-1G 368199-5G
SIAL368199-1GEA 32.6 GBP
SIAL368199-1G SIAL368199-5G
(S)-(-)-α,α-Diphenylprolinol, Sigma-Aldrich®
(S)-(-)-α,α-Diphenylprolinol
Formula: C₁₇H₁₉NO
MW: 253.35 g/mol
MDL Number: MFCD00075506
CAS Number: 112068-01-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR