3-Chloro-2-(2-bromoacetyl)thiophene

Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: 2-Bromo-1-(3-chlorothiophen-2-yl)ethan-1-one

9J-909.500 9J-909.1 9J-909.5 9J-909.10
BIOT9J-909.500EA 0 GBP
BIOT9J-909.500 BIOT9J-909.1 BIOT9J-909.5 BIOT9J-909.10
3-Chloro-2-(2-bromoacetyl)thiophene
3-Chloro-2-(2-bromoacetyl)thiophene
Formula: C₆H₄BrClOS
MDL Number: MFCD00793892
CAS Number: 256486-71-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR