2,6-Dimethoxyaniline hydrochloride

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

AN-1245-5G AN-1245-1G
COBBAN-1245-5GEA 0 GBP
COBBAN-1245-5G COBBAN-1245-1G
2,6-Dimethoxyaniline hydrochloride
2,6-Dimethoxyaniline hydrochloride
Formula: C₈H₁₂ClNO₂
MW: 189.6394 g/mol
MDL Number: MFCD09878313
CAS Number: 375397-36-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR