3-Bromo-5-chloro-4-methoxyaniline

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

AN-1562-5G AN-1562-1G
COBBAN-1562-5GEA 0 GBP
COBBAN-1562-5G COBBAN-1562-1G
3-Bromo-5-chloro-4-methoxyaniline
3-Bromo-5-chloro-4-methoxyaniline
Formula: C₇H₇BrClNO
MDL Number: MFCD15143449
CAS Number: 84972-58-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR