1-(Trimethylsilyl)-1,2,4-triazole

Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: 1-(Trimethylsilyl)-1H-1,2,4-triazole

SILS20325.5 SILS20325.25
SILNSILS20325.5EA 0 GBP
SILNSILS20325.5 SILNSILS20325.25
1-(Trimethylsilyl)-1,2,4-triazole
1-(Trimethylsilyl)-1,2,4-triazole
Formula: C₅H₁₁N₃Si
MW: 141.25 g/mol
MDL Number: MFCD00046070
CAS Number: 18293-54-4
EINECS: 242-170-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR