3,4-Difluoro-2-methoxyaniline

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

AN-1122-5G AN-1122-1G
COBBAN-1122-5GEA 0 GBP
COBBAN-1122-5G COBBAN-1122-1G
3,4-Difluoro-2-methoxyaniline
3,4-Difluoro-2-methoxyaniline
Formula: C₇H₇F₂NO
MW: 159.133 g/mol
MDL Number: MFCD07368889
CAS Number: 114076-35-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR