(±)-3-Hydroxytetrahydrofuran
Supplier: FLUOROCHEM LTD (CHEMPUR)
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
FL5327.5
FL5327.25
005327-100G
005327-25G
CHMPFL5327.5EA
0
GBP
CHMPFL5327.5
CHMPFL5327.25
FLUO005327-100G
FLUO005327-25G
(±)-3-Hydroxytetrahydrofuran
(±)-3-Hydroxytetrahydrofuran
Formula:
C₄H₈O₂ |
MDL Number:
MFCD00005374 CAS Number: 453-20-3 EINECS: 207-219-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...