2-Amino-6-chloropyrazine

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

002753-1G 002753-5G 002753-25G
FLUO002753-1GEA 0 GBP
FLUO002753-1G FLUO002753-5G FLUO002753-25G
2-Amino-6-chloropyrazine
2-Amino-6-chloropyrazine
Formula: C₄H₄ClN₃
MW: 129.55 g/mol
MDL Number: MFCD00055024
CAS Number: 33332-28-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR