Oxindole-6-boronic acid
Supplier: COMBI-BLOCKS
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synonyms:
(2-Oxoindolin-6-yl)boronic acid
BB-5414-1G
BB-5414-250M
COBBBB-5414-1GEA
0
GBP
COBBBB-5414-1G
COBBBB-5414-250M
Oxindole-6-boronic acid
Oxindole-6-boronic acid
Formula:
C₈H₈BNO₃ MW: 176.9650 g/mol |
MDL Number:
MFCD12913936 CAS Number: 1217500-61-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...