4-Pentylcyclohexanone

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

OS-7417-5G OS-7417-25G OS-7417-100G
COBBOS-7417-5GEA 0 GBP
COBBOS-7417-5G COBBOS-7417-25G COBBOS-7417-100G
4-Pentylcyclohexanone
4-Pentylcyclohexanone
Formula: C₁₁H₂₀O
MW: 168.28 g/mol
MDL Number: MFCD00059581
CAS Number: 61203-83-6
EINECS: 406-670-4
UN: 3082
ADR: 9,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR