3-(4-Hydroxyphenyl)propiononitrile

Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: 3-(4-Hydroxyphenyl)propionitrile

H2330.10 H2330.50
XXXXH2330.10EA 0 GBP
XXXXH2330.10 XXXXH2330.50
3-(4-Hydroxyphenyl)propiononitrile
3-(4-Hydroxyphenyl)propiononitrile
Formula: C₉H₉NO
MW: 147.18 g/mol
MDL Number: MFCD00016492
CAS Number: 17362-17-3
EINECS: 241-393-0
UN: 3439
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR