2-Acetyl-4,5-dibromothiophene
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
9Y-0839.10
9Y-0839.1
9Y-0839.5
9Y-0839.500
BIOT9Y-0839.10EA
0
GBP
BIOT9Y-0839.10
BIOT9Y-0839.1
BIOT9Y-0839.5
BIOT9Y-0839.500
2-Acetyl-4,5-dibromothiophene
2-Acetyl-4,5-dibromothiophene
Formula:
C₆H₄Br₂OS |
CAS Number:
7209-12-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...