Benzo[j]fluoranthene
Supplier: LGC Standards PROMOCHEM
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
PROOCILULM241125EA
997
GBP
PROOCILULM241125
Benzo[j]fluoranthene
Benzo[j]fluoranthene
Formula:
C₂₀H₁₂ |
CAS Number:
205-82-3 EINECS: 205-910-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...