Indomethacin
Supplier: Molekula
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
MOLE62725591-25GEA
0
GBP
MOLE62725591-25G
Indomethacin
Indomethacin
Formula:
C₁₉H₁₆ClNO₄ MW: 357.80 g/mol |
MDL Number:
MFCD00057095 CAS Number: 53-86-1 EINECS: 200-186-5 UN: 2811 ADR: 6.1,I |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...