Indomethacin

Supplier: EHRENSTORFER GMBH
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

EHERC14325000EA 109 GBP
EHERC14325000
Indomethacin
Indomethacin
Formula: C₁₉H₁₆ClNO₄
MW: 357.80 g/mol
MDL Number: MFCD00057095
CAS Number: 53-86-1
EINECS: 200-186-5
UN: 2811
ADR: 6.1,I

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR