Alcian blue 8GX

Supplier: TSC BIOSCIENCES LTD
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Warning

TCSBHD1040EA 177 GBP
TCSBHD1040
Alcian blue 8GX
Alcian blue 8 GX
Formula: C₅₆H₆₈Cl₄CuN₁₆S₄
MW: 1298.86 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00010720
CAS Number: 33864-99-2
EINECS: 251-705-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR