2,3-Dihydrobenzo[b]furan, Sigma-Aldrich®
Supplier: Merck
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
M-Clarity™ quality level = MQ100
183962-25G
183962-5G
SIAL183962-25GEA
111
GBP
SIAL183962-25G
SIAL183962-5G
2,3-Dihydrobenzo[b]furan, Sigma-Aldrich®
2,3-Dihydrobenzo[b]furan
Formula:
C₈H₈O MW: 120.15 g/mol |
MDL Number:
MFCD00005855 CAS Number: 496-16-2 EINECS: 207-817-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...