2,3-Dihydrobenzo[b]furan
Supplier: FLUOROCHEM
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
CFF699.25
017117-1G
017117-100G
017117-25G
017117-5G
CHMPCFF699.25EA
0
GBP
CHMPCFF699.25
FLUO017117-1G
FLUO017117-100G
FLUO017117-25G
FLUO017117-5G
2,3-Dihydrobenzo[b]furan
2,3-Dihydrobenzo[b]furan
Formula:
C₈H₈O MW: 120.15 g/mol |
MDL Number:
MFCD00005855 CAS Number: 496-16-2 EINECS: 207-817-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...