2,3-Dihydrobenzo[b]furan

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

OS-7551-100G OS-7551-25G OS-7551-5G
COBBOS-7551-100GEA 0 GBP
COBBOS-7551-100G COBBOS-7551-25G COBBOS-7551-5G
2,3-Dihydrobenzo[b]furan
2,3-Dihydrobenzo[b]furan
Formula: C₈H₈O
MW: 120.15 g/mol
MDL Number: MFCD00005855
CAS Number: 496-16-2
EINECS: 207-817-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR