1-Methylimidazole-2-carboxylic acid, Sigma-Aldrich®

Supplier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: 1-Methyl-1H-imidazole-2-carboxylic acid

M-Clarity™ quality level = MQ100

SIAL576220-1GEA 58.6 GBP
SIAL576220-1G
1-Methylimidazole-2-carboxylic acid, Sigma-Aldrich®
1-Methylimidazole-2-carboxylic acid
Formula: C₅H₆N₂O₂
MW: 126.11 g/mol
MDL Number: MFCD01863421
CAS Number: 20485-43-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR