Imidazole-4-carboxylic acid ethyl ester

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: Ethyl imidazole-4-carboxylate

HI-1187-1G HI-1187-25G HI-1187-5G
COBBHI-1187-1GEA 0 GBP
COBBHI-1187-1G COBBHI-1187-25G COBBHI-1187-5G
Imidazole-4-carboxylic acid ethyl ester
Imidazole-4-carboxylic acid ethyl ester
Formula: C₆H₈N₂O₂
MW: 140.14 g/mol
MDL Number: MFCD07363812
CAS Number: 23785-21-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR