Hymecromone sodium salt (4-methylumbelliferone sodium salt, 7-hydroxy-4-methylcoumarin sodium salt), Sigma-Aldrich®

Supplier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5


M-Clarity™ quality level = MQ200

M1508-10G M1508-100G M1508-25G
SIALM1508-10GEA 98.3 GBP
SIALM1508-10G SIALM1508-100G SIALM1508-25G
Hymecromone sodium salt (4-methylumbelliferone sodium salt, 7-hydroxy-4-methylcoumarin sodium salt), Sigma-Aldrich®
Hymecromone sodium salt (4-Methylumbelliferone sodium salt, 7-Hydroxy-4-methylcoumarin sodium salt)
Formula: C₁₀H₇NaO₃
CAS Number: 5980-33-6
EINECS: 227-790-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR