6-Methoxy-2H-chromene-3-carboxylic acid

Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

33-0040.1 33-0040.5
CHMP33-0040.1EA 0 GBP
CHMP33-0040.1 CHMP33-0040.5
6-Methoxy-2H-chromene-3-carboxylic acid
6-Methoxy-2H-chromene-3-carboxylic acid
Formula: C₁₁H₁₀O₄
MW: 206.1947 g/mol
MDL Number: MFCD00202057
CAS Number: 57543-62-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR