6-Methoxy-2H-chromene-3-carboxylic acid methyl ester

Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: Methyl 6-methoxy-2H-chromene-3-carboxylate

33-0042.1 33-0042.5
CHMP33-0042.1EA 0 GBP
CHMP33-0042.1 CHMP33-0042.5
6-Methoxy-2H-chromene-3-carboxylic acid methyl ester
6-Methoxy-2H-chromene-3-carboxylic acid methyl ester
Formula: C₁₂H₁₂O₄
MDL Number: MFCD00202061
CAS Number: 338759-76-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR