7-Methoxy-2H-chromene-3-carboxylic acid methyl ester

Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

33-0048.1 33-0048.5
CHMP33-0048.1EA 0 GBP
CHMP33-0048.1 CHMP33-0048.5
7-Methoxy-2H-chromene-3-carboxylic acid methyl ester
7-Methoxy-2H-chromene-3-carboxylic acid methyl ester
Formula: C₁₂H₁₂O₄
MDL Number: MFCD04114606
CAS Number: 86236-08-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR