2,2-Dimethylsuccinic anhydride

Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: 2,2-Dimethylsuccinicanhydride

D5800.5 D5800.25
XXXXD5800.5EA 0 GBP
XXXXD5800.5 XXXXD5800.25
2,2-Dimethylsuccinic anhydride
2,2-Dimethylsuccinic anhydride
Formula: C₆H₈O₃
MW: 128.1259 g/mol
MDL Number: MFCD00022608
CAS Number: 17347-61-4
EINECS: 241-371-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR