2,2-Dimethylsuccinic anhydride
Supplier: FLUOROCHEM
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synonyms:
2,2-Dimethylsuccinicanhydride
002557-1G
002557-5G
FLUO002557-1GEA
0
GBP
FLUO002557-1G
FLUO002557-5G
2,2-Dimethylsuccinic anhydride
2,2-Dimethylsuccinic anhydride
Formula:
C₆H₈O₃ MW: 128.1259 g/mol |
MDL Number:
MFCD00022608 CAS Number: 17347-61-4 EINECS: 241-371-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...