4,4,4-Trifluoro-2-butanone

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

007508-5G 007508-25G 007508-1G
FLUO007508-5GEA 0 GBP
FLUO007508-5G FLUO007508-25G FLUO007508-1G
4,4,4-Trifluoro-2-butanone
4,4,4-Trifluoro-2-butanone
Formula: C₄H₅F₃O
MW: 126.08 g/mol
MDL Number: MFCD00077601
CAS Number: 2366-70-3
UN: 1224
ADR: 3,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR