4,4,4-Trifluoro-2-butanone ≥97%
Supplier: Thermo Scientific
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Warning
|
B21514.03
B21514.06
B21514.03EA
32.7
GBP
B21514.03
B21514.06
4,4,4-Trifluoro-2-butanone ≥97%
4,4,4-Trifluoro-2-butanone
Purity: 97%
CAS No.: 2366-70-3
Molecular Formula: CF3CH2COCH3
Formula Weight: 126.08
Flash Point: 37°(99°F)
Boiling Point: 95-96°
UN No.: 1224
MDL No.: MFCD00077601
Formula:
C₄H₅F₃O MW: 126.08 g/mol Boiling Pt: 95…96 °C Density: 1.05 Flash Pt: 37 °C (99 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00077601 CAS Number: 2366-70-3 UN: 1224 ADR: 3,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...