3-Chloro-2-iodo-1,1,1-trifluoropropane
Supplier: FLUOROCHEM
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
010273-5G
010273-25G
FLUO010273-5GEA
0
GBP
FLUO010273-5G
FLUO010273-25G
3-Chloro-2-iodo-1,1,1-trifluoropropane
3-Chloro-2-iodo-1,1,1-trifluoropropane
Formula:
C₃H₃ClF₃I |
MDL Number:
MFCD01862016 CAS Number: 113402-77-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...