2-Iodonicotinonitrile

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

CA-4223-5G CA-4223-1G
COBBCA-4223-5GEA 0 GBP
COBBCA-4223-5G COBBCA-4223-1G
2-Iodonicotinonitrile
2-Iodonicotinonitrile
Formula: C₆H₃IN₂
MDL Number: MFCD08436053
CAS Number: 490039-73-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR