5-Amino-2-chlorophenol
Supplier: COMBI-BLOCKS
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synonyms:
2-Chloro-5-aminophenol
AN-1204-25G
AN-1204-5G
COBBAN-1204-25GEA
0
GBP
COBBAN-1204-25G
COBBAN-1204-5G
5-Amino-2-chlorophenol
5-Amino-2-chlorophenol
Formula:
C₆H₆ClNO MW: 143.57 g/mol |
MDL Number:
MFCD01707858 CAS Number: 6358-06-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...