Bis(2-bromoethyl)ether
Supplier: FLUOROCHEM
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synonyms:
1-Bromo-2-(2-bromoethoxy)ethane, 2-Bromoethyl ether
005451-25G
005451-100G
FLUO005451-25GEA
0
GBP
FLUO005451-25G
FLUO005451-100G
Bis(2-bromoethyl)ether
Bis(2-bromoethyl) ether
Formula:
C₄H₈Br₂O MW: 231.93 g/mol |
MDL Number:
MFCD00039196 CAS Number: 5414-19-7 EINECS: 226-504-2 UN: 2810 ADR: 6.1,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...