Flash chromatography columns, PuriFlash®, NH₂ bonded silica
Supplier: Interchim
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
PuriFlash®
INTEPF30NH2/6GEA 112 GBP
INTEPF30NH2/6G INTEPF30NH2/35G INTEPF50NH2/1250G INTEPF30NH2/55G INTEPF50NH2/20G INTEPF50NH2/55G INTEPF50NH2/35G INTEPF50NH2/450G INTEPF30NH2/20G INTEPF30NH2/1250G INTEPF50NH2/120G INTEPF50NH2/175G INTEPF50NH2/300G INTEPF30NH2/120G INTEPF30NH2/175G INTEPF30NH2/2500G INTEPF30NH2/450G INTEPF50NH2/2500G INTEPF50NH2/6G INTEPF30NH2/300G
Flash chromatography columns, PuriFlash®, NH₂ bonded silica
Chromatography Columns
These flash chromatography columns with NH₂ bonded silica are used for polar amine compounds with reduced amine-silanol interaction and are also suitable for carbohydrates. They eliminate the use of TEA (triethylamine) for basic compounds.
  • Base material is high performance spherical silica
  • pH stability ranges from 2 to 6,5