Flash chromatography columns, PuriFlash®, DEAP bonded silica
Supplier: Interchim
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
PuriFlash®
INTEPF50DEAP/175GEA 392 GBP
INTEPF50DEAP/175G INTEPF50DEAP/120G INTEPF50DEAP/450G INTEPF50DEAP/20G INTEPF50DEAP/1250G INTEPF50DEAP/35G INTEPF50DEAP/6G INTEPF50DEAP/55G INTEPF50DEAP/2500G INTEPF50DEAP/300G
Flash chromatography columns, PuriFlash®, DEAP bonded silica
Chromatography Columns
These flash chromatography columns with DEAP (DiethylAminoPropyl) bonded silica are suitable for charged polar organics compounds. Compatible with MeOH and water for use in normal or reverse phase.
  • Base material is granular silica