1,3-Bis(4-chlorophenyl)imidazolium chloride ≥97%

Supplier: Strem Chemicals, Inc.
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Warning

Synonyms: 1,3-Bis(4-chlorophenyl)-1H-imidazol-3-ium chloride

STRE07-0490-250MGEA 0 GBP
STRE07-0490-250MG
1,3-Bis(4-chlorophenyl)imidazolium chloride ≥97%
1,3-Bis(4-chlorophenyl)imidazolium chloride

NHC

Formula: C₁₅H₁₁Cl₃N₂
MW: 325.620 g/mol
Melting Pt: 251…252 °C
MDL Number: MFCD03788924
CAS Number: 141556-46-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR