ABTS (Diammonium 2,2'-azinobis[3-ethyl-2,3-dihydrobenzothiazole-6-sulphonate]), Sigma-Aldrich®
Supplier: Merck Millipore (Oncogene)
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Warning
|
Synonyms:
2,2′-Azino-bis(3-ethylbenzthiazoline-6-sulphonic acid) diammonium salt
, Ammonium 2,2'-(hydrazine-1,2-diylidene)bis(3-ethyl-2,3-dihydrobenzo[d]thiazole-6-sulphonate)
, ABTS
194437-1EA
160
GBP
194437-1
ABTS (Diammonium 2,2'-azinobis[3-ethyl-2,3-dihydrobenzothiazole-6-sulphonate]), Sigma-Aldrich®
ABTS (Diammonium 2,2'-azinobis[3-ethyl-2,3-dihydrobenzothiazole-6-sulphonate])
Formula:
C₁₈H₂₄N₆O₆S₄ MW: 548.68 g/mol Storage Temperature: Refrigerator |
MDL Number:
MFCD00010404 CAS Number: 30931-67-0 EINECS: 250-396-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...