Acridine orange (hemizinc salt)

Supplier: Serva
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Danger

Synonyms: Acridine orange hemi(Zinc chloride) saltAcridine orangeBasic orange 143,6-Bis(dimethylamino)acridine hydrochloride zinc chloride double saltEuchrysine 3RXEuchrysin GNYN,N,N′,N′-Tetramethylacridine-3,6-diamine monohydrochloride compd. with zinc dichloride3,6-Bis(dimethylamino)acridinium chloride hemi(zinc chloride salt)

SERA10665.02EA 0 GBP
SERA10665.02
Acridine orange (hemizinc salt)
Acridine orange (hemizinc salt)
Formula: C₁₇H₂₀ClN₃·HCl·½ZnCl₂
MW: 369.96 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00081043
CAS Number: 10127-02-3
EINECS: 233-353-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR