(+)-Di-p-toluoyl-D-tartaric acid monohydrate

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: (2S,3S)-2,3-Bis((4-methylbenzoyl)oxy)succinic acid hydrate, Di-p-toluoyl-D-tartaric acid monohydrate

011667-100G 011667-25G
FLUO011667-100GEA 0 GBP
FLUO011667-100G FLUO011667-25G
(+)-Di-p-toluoyl-D-tartaric acid monohydrate
(+)-Di-p-toluoyl-D-tartaric acid monohydrate
Formula: C₂₀H₁₈O₈·1H₂O
MW: 386.358 g/mol (anhy)
MDL Number: MFCD00149567
CAS Number: 71607-31-3
EINECS: 251-132-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR