1-Naphthyl phosphate

Supplier: Molekula
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

MOLE66442790-5GEA 0 GBP
MOLE66442790-5G
1-Naphthyl phosphate
1-Naphthyl phosphate
Formula: C₁₀H₉O₄P
CAS Number: 1136-89-6
EINECS: 214-502-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR