4-Nitroquinoline-N-oxide

Supplier: EHRENSTORFER GMBH
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: 4-Nitroquinoline 1-oxide

EHERC15558000EA 96.8 GBP
EHERC15558000
4-Nitroquinoline-N-oxide
4-Nitroquinoline N-oxide
Formula: C₉H₆N₂O₃
MW: 190.1555 g/mol
MDL Number: MFCD00006738
CAS Number: 56-57-5
EINECS: 200-281-1
UN: 2811
ADR: 6.1,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR